Recombinant Mouse PDGFA (from E. coli)
Catalog # ABNOP7478.10UG
Supplier: Abnova
CAS Number:
Specifications
- Cdt:10 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Mouse
- ID de gène:P20033
- Nom de la protéine/du peptide:PDGFA
- Sequence:87a.a.-211a.a.