Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Human CCL25 (from E. coli)
Recombinant Human CCL25 (from E. coli)
Catalog # ABNOP7223.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CCL25 (from E. coli)
Catalog # ABNOP7223.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Cdt:
    20 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • ID de gène:
    O15444
  • Nom de la protéine/du peptide:
    CCL25
  • Sequence:
    24a.a.-150a.a.

Specifications

About this item