Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Mouse CCL20 (from E. coli)
Recombinant Mouse CCL20 (from E. coli)
Catalog # ABNOP7287.5UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse CCL20 (from E. coli)
Catalog # ABNOP7287.5UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Cdt:
    5 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • ID de gène:
    O89093
  • Nom de la protéine/du peptide:
    CCL20
  • Sequence:
    28a.a.-97a.a.

Specifications

About this item