0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Protéines et Peptides
Recombinant Mouse EGF (from E. coli)
Recombinant Mouse EGF (from E. coli)
ABNOP7299.10UG
:
Abnova
:
Recombinant Mouse EGF (from E. coli)
ABNOP7299.10UG
:
Abnova
:
Cdt:
10 µG
Type de protéine/peptide:
Recombinant
Source:
E. coli
Species:
Mouse
ID de gène:
P01132
Nom de la protéine/du peptide:
EGF
Sequence:
977a.a.-1029a.a.