Anti-TNFRSF9 Mouse Recombinant Antibody [clone: 4B4-1-1]
Catalog # ABNORAB03645.200UG
Supplier: Abnova
Specifications
- Symbole de l'antigène:TNFRSF9
- Clonalité:Recombinant
- Clone:4B4-1-1
- Hôte:Mouse
- Réactivité:Human
- Immunogène:Recombinant mouse monoclonal antibody raised against human TNFRSF9.
- Cdt:200 µG