Anti-Angiopoietin 1 Rabbit Recombinant Antibody [clone: R06-1F9]
Catalog # ABNORAB01740.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:Angiopoietin 1
- Clonalité:Recombinant
- Clone:R06-1F9
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Angiopoietin 1
- Cdt:100 µl