Anti-ATM Rabbit Recombinant Antibody [clone: R08-8H3]
Catalog # ABNORAB01710.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:ATM
- Clonalité:Recombinant
- Clone:R08-8H3
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human ATM
- Cdt:100 µl