Anti-Profilin 1 Rabbit Recombinant Antibody [clone: R06-6H7]
ABNORAB01694.100UL
: Abnova
- Symbole de l'antigène:Profilin 1
- Clonalité:Recombinant
- Clone:R06-6H7
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Profilin 1
- Cdt:100 µl