Order Entry
Switzerland
ContactUsLinkComponent
Anti-POSTN Rabbit Recombinant Antibody [clone: R03-7E3]
Anti-POSTN Rabbit Recombinant Antibody [clone: R03-7E3]
Catalog # ABNORAB02012.100UL
Supplier:  Abnova
Anti-POSTN Rabbit Recombinant Antibody [clone: R03-7E3]
Catalog # ABNORAB02012.100UL
Supplier:  Abnova

Specifications

  • Symbole de l'antigène:
    POSTN
  • Clonalité:
    Recombinant
  • Clone:
    R03-7E3
  • Hôte:
    Rabbit
  • Réactivité:
    Human
  • Immunogène:
    Recombinant rabbit monoclonal antibody raised against human POSTN.
  • Cdt:
    100 µl

Specifications

About this item