Anti-RAB10 Rabbit Recombinant Antibody [clone: R06-2A3]
Catalog # ABNORAB01969.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:RAB10
- Clonalité:Recombinant
- Clone:R06-2A3
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human RAB10.
- Cdt:100 µl