Anti-H4C1 Rabbit Recombinant Antibody [clone: R05-8H9]
Codice catalogo ABNORAB02219.100UL
Nome del fornitore: Abnova
Specifiche
- Symbole de l'antigène:H4C1
- Clonalité:Recombinant
- Clone:R05-8H9
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human H4C1.
- Cdt:100 µl