Anti-GNA13 Rabbit Recombinant Antibody [clone: R08-4F2]
Catalog # ABNORAB02270.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:GNA13
- Clonalité:Recombinant
- Clone:R08-4F2
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human GNA13.
- Cdt:100 µl