Anti-FLI1 Rabbit Recombinant Antibody [clone: R01-0C2]
Catalog # ABNORAB02277.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:FLI1
- Clonalité:Recombinant
- Clone:R01-0C2
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human FLI1.
- Cdt:100 µl