Anti-EPN1 Rabbit Recombinant Antibody [clone: R02-2D1]
Catalog # ABNORAB02295.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:EPN1
- Clonalité:Recombinant
- Clone:R02-2D1
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human EPN1.
- Cdt:100 µl