Anti-MAPK3 Rabbit Recombinant Antibody [clone: R08-3C6]
Catalog # ABNORAB02294.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:MAPK3
- Clonalité:Recombinant
- Clone:R08-3C6
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human MAPK3.
- Cdt:100 µl