Anti-GOT2 Rabbit Recombinant Antibody [clone: R09-9G8]
Catalog # ABNORAB02290.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:GOT2
- Clonalité:Recombinant
- Clone:R09-9G8
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human GOT2.
- Cdt:100 µl