Anti-FH Rabbit Recombinant Antibody [clone: R08-3F2]
Catalog # ABNORAB02281.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:FH
- Clonalité:Recombinant
- Clone:R08-3F2
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human FH.
- Cdt:100 µl