Anti-FGF1 Rabbit Recombinant Antibody [clone: R05-6A4]
Catalog # ABNORAB02282.100UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:FGF1
- Clonalité:Recombinant
- Clone:R05-6A4
- Hôte:Rabbit
- Réactivité:Human
- Immunogène:Recombinant rabbit monoclonal antibody raised against human FGF1.
- Cdt:100 µl