Order Entry
Switzerland
ContactUsLinkComponent
Anti-MEF2A Rabbit Recombinant Antibody [clone: R06-9A6]
Anti-MEF2A Rabbit Recombinant Antibody [clone: R06-9A6]
Catalog # ABNORAB02110.100UL
Supplier:  Abnova
Anti-MEF2A Rabbit Recombinant Antibody [clone: R06-9A6]
Catalog # ABNORAB02110.100UL
Supplier:  Abnova

Specifications

  • Symbole de l'antigène:
    MEF2A
  • Clonalité:
    Recombinant
  • Clone:
    R06-9A6
  • Hôte:
    Rabbit
  • Réactivité:
    Human
  • Immunogène:
    Recombinant rabbit monoclonal antibody raised against human MEF2A.
  • Cdt:
    100 µl

Specifications

About this item