Anti-LMNA (pSer22) Rabbit Monoclonal Antibody [clone: CF12] ()
Catalog # ABNOMAB23550.200UL
Supplier: Abnova
Specifications
- Symbole de l'antigène:LMNA
- Clonalité:Monoclonal
- Clone:CF12
- Hôte:Rabbit
- Réactivité:Human
- Modification:pSer22
- Immunogène:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human LMNA.
- Cdt:200 µl