Anti-MTOR (pSer2448) Rabbit Monoclonal Antibody [clone: E11] (FITC)
Catalog # ABNOMAB23511.100
Supplier: Abnova
Specifications
- Symbole de l'antigène:MTOR
- Clonalité:Monoclonal
- Clone:E11
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- Réactivité:Human
- Modification:pSer2448
- Immunogène:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human MTOR.
- Cdt:100 AU