Order Entry
Switzerland
ContactUsLinkComponent
Anti-MTOR (pSer2448) Rabbit Monoclonal Antibody [clone: E11] (APC)
Anti-MTOR (pSer2448) Rabbit Monoclonal Antibody [clone: E11] (APC)
Catalog # ABNOMAB23512.100
Supplier:  Abnova
Anti-MTOR (pSer2448) Rabbit Monoclonal Antibody [clone: E11] (APC)
Catalog # ABNOMAB23512.100
Supplier:  Abnova

Specifications

  • Symbole de l'antigène:
    MTOR
  • Clonalité:
    Monoclonal
  • Clone:
    E11
  • Conjugaison:
    APC (Allophycocyanin)
  • Hôte:
    Rabbit
  • Réactivité:
    Human
  • Modification:
    pSer2448
  • Immunogène:
    Rabbit monoclonal antibody raised against synthetic phosphopeptide of human MTOR.
  • Cdt:
    100 AU

Specifications

About this item