ToughStripe® Arrows
BRDY175655
: Brady
- Désignation:ToughStripe® arrows
- Dimensions:101,6×254 mm
- Matière:Polyester
- Cdt:25
ToughStripe® Arrows provide visible floor marking for improved safety and organisation in almost any industrial environment.
- Polyester with polyester overlaminate
- Permanent rubber adhesive
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Ideal for marking aisles, passageways and storage locations, and identifying equipment and egress paths
- Can be easily applied by one person working alone
Extremely durable ToughStripe® die-cut floor marking arrows are made of rugged polyester with ultra-aggressive adhesive and hold up against forklift traffic.
: 25 labels per pack.