LCSM (Lin's Cupric Sulphate Medium)
LCSM (Lin's Cupric Sulphate Medium)
LCSM (Lin's Cupric Sulphate Medium), 260 g, Plastic bottle, 1
N° de catalogue DOEH923552244
Fournisseur:   DOEHLER GROUP
LCSM (Lin's Cupric Sulphate Medium)
N° de catalogue DOEH923552244
Fournisseur:  DOEHLER GROUP
Numéro CAS:  
LCSM (Lin's Cupric Sulphate Medium)
LCSM (Lin's Cupric Sulphate Medium)
LCSM (Lin's Cupric Sulphate Medium), 260 g, Plastic bottle, 1
Numéro CAS:  
Prix
SFr. 307.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Siebel-Instituts agar medium for the selective detection and quantitative determination of NON-Saccharomyces wild yeast populations in samples from the brewing process.
  • Certified by the Siebel-Institute for technology
  • Ready-to-use powder with batch-adjusted cupric sulfate solution
  • Functionality tested with strains originally isolated from the brewing environment
The specially developed nutrient composition, the media primarily enhance the growth of NON-Saccharomyces wild yeasts, whereas the normal brewing culture is mainly inhibited by the addition of cupric sulfate solution. LCSM can be applied to filterable samples (e.g. beer and rinsing water samples) but also in brewing culture yeast with high cell counts.
Documents
Spécifications
Spécifications
  • Type de conditionnement:
    Plastic bottle
  • Cdt:
    1
  • Désignation:
    LCSM (Lin's Cupric Sulphate Medium)
  • Remplissage:
    260 g