- Désignation:Wi-Fi streamer with operating manual, Dimension: 36W×58L×32H, Frequency: 2,412 - 2,485 GHz, Weight: 57 g
- Cdt:1
Coupling with Dino-Lite AF series, the WF-20 is a Wi-Fi streamer adding further versatility, flexibility, and agility to Dino-Lite microscope. Thanks to stable image transmission and long battery life (more than 2,5 hours), the WF-20 is well suited but not limited to field applications. The WF-20 can also be configured as a wireless router for staying connected with internet or a network during usage.
- Interface release button
- Wi-Fi signal LED indicator
- Battery LED indicator
- On/off power button
- Reset pinhole button
- Mini USB port (for charging purpose)
The WF-20 allows AF microscope models to be used wirelessly in connection with an iOS (iPhone/iPad) or Android app on any tablet, smartphone or even in the DinoCapture 2.0 software for windows computers. The WiFi streamer is ideal for field work or presentations and can stream from one Dino-Lite to multiple devices. When using the iOS app or DinoCapture, users can perform measurements and take advantage of the automatic magnification reading (AMR) with compatible Dino-Lite AMR and measurement models. The WF-20 can be used anywhere to create a WiFi signal for streaming the Dino-Lite live images.
Compatible with: Dino-Lite Edge AF series, AF4x15 series.
Please note: The free DinoConnect app needs to be downloaded from the app store or google play.