Specifications
- Désignation:THUNDER image live cell
- Cdt:1
Specifications
About this item
THUNDER Imager is designed to meet the needs of tomorrow's 3D cell culture applications. Work effortlessly with modern sample carrier formats, such as 8-chamber slides or 96-well plates. Generate more data in less time to boost the statistical power of your assay. Get closer to observing the real physiology of your 3D specimens by minimizing light exposure and obtaining brilliant images, while enjoying great ease-of-use. THUNDER imager 3D Live cell, 3D cell culture and 3D assay decode 3D biology in real time.
- High throughput for better statistics and efficiency
- High imaging performance from an ease-to-use instruments
- Optimal physiological conditions for meaningful results
With a THUNDER Imager 3D Live Cell, 3D Cell Culture and 3D Assay you take full advantage of:
High-speed positioning with the quantum stage and synapse real-time control (Quantum Stage only available with 3DCC/3DLC) High-speed illumination with a multi-line LED light source (Multiline LED light source optional for 3D assay)
High-sensitivity thanks to SCMOS technology and Leica optics
Efficient removal of out-of-focus blur even from single plane acquisitions
Great ease-of-use, less training time
Compatible with multi-well carriers for automated water immersion (optional) to attain better sensitivity and resolutionCONNECTWITH USI