96 Well Plate, RCH CEREX MAESTRO
Catalog # TCAT613-0151RCH
Supplier: Tecan
Specifications
- Nb de puits:96
- Désignation:96-well plate for RCH CEREX MAESTRO 1 ml
- Greffage:MAESTRO
- Remplissage:15 mg
- Cdt:1
Specifications
About this item
Silica-based sorbent with C18 bonding in proprietary coverage.
- Reversed-phased
- Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.