Microfibre Cart
Catalog # 129-1947
Supplier: Rubbermaid Commercial Products
Specifications
- Couleur:noir
- l×p×h:558×1225×1117 mm
- Cdt:1
Specifications
About this item
The Rubbermaid Commercial high-capacity janitorial cleaning cart with bins is a complete system solution for your cleaning needs. Flexible and configurable to fit your ever-changing cleaning needs.
- 40% more storage space than traditional carts minimises trips for additional supplies and maximises staff productivity
- Non-marking 10,2 cm casters provide maximum maneuverability
- 28,4 L removable bins offer more organization and the ability to prestock cleaning supplies for the upcoming shift
- Interchangeable handles and removable platform allow user to easily expand or reduce the footprint of the cart to meet specific cleaning and storage needs
- Rubber tool grips and hooks securely hold up to five stick goods, while hooks hold up to four safety signs
- Compact size fits great in space-constrained areas; with a high capacity-to-footprint ratio, only 55,9 cm wide
- Mop bucket platform with securing grooves keeps WaveBrake® or microfiber charging buckets securely attached to cart
Expansive storage space with removable bins and caddies let staff restock before they clean, increasing productivity by eliminating search time for additional supplies.
Delivery: Includes 4 colour-coded buckets 5 L, 2 yellow organising boxes 28 L, 1 waste cover and storage compartment and 1 yellow high capacity vinyl bag 92 L.