Order Entry
Switzerland
ContactUsLinkComponent
Multiskan™ FC Microplate Photometer
Multiskan™ FC Microplate Photometer
  HERA51119000
undefined
Multiskan™ FC Microplate Photometer
  HERA51119000

 

  • Longueur d'onde:
    340 - 850 nm
  • Détecteur:
    Photodetector
  • Source de lumière:
    Quartz halogen lamp
  • Types de plaques:
    96-well plate
  • Résolution:
    0,001 Abs
  • Tension:
    100 - 240 V
  • Sélection de la longueur d'onde:
    Filters
  • Désignation:
    Multiskan FC
  • Exigence électrique:
    100/240 V, 50/60 Hz
  • Gamme de longueur d'onde:
    340 - 850 nm
  • Précision d'absorbance:
    ±1% (0 to 3 Abs) or ±0,003 Abs, which ever is greater at 405 nm
  • Filtres:
    8-position filter wheel; filters installed: 405 nm, 450 nm and 620 nm
  • Linéarité:
    0 - 3 Abs, ±2% at 405 nm
  • Mode de mesure:
    Absorbance
  • Durée de mesure:
    7 s for 96-well plate (Fast mode), 13 s for 96-well plate (Normal mode)
  • Précision:
    CV ≤0,2% (0,3 - 3 Abs) at 405 nm
  • Agitation:
    Linear
  • Logiciel:
    SkanIt Software
  • L×P×H:
    290×400×220 mm
  • Cdt:
    1

 

 

The Multiskan FC has a 340 to 850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard.

  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50 °C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments

A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.

A broad wavelength range of 340 to 850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves. Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 and 620 nm, preinstalled.