Scotch-Weld™ DP190 Epoxy Adhesive Grey
Catalog # MMMA190.50
Supplier: 3M
Specifications
- Volume:48,5 ml
- Couleur:gris
- Désignation:Scotch-Weld™ Epoxy adhesive, DP190
- Cdt:12
Specifications
About this item
Scotch-Weld™ Epoxy adhesive DP190 is a two-part epoxy adhesive that delivers exceptional performance with high shear and peel strength. Extremely compatible, it is formulated to bond to a wide range of materials, including metals, ceramics, wood, fibreboard, glass, rubber, and many plastics.
- Apply with ease with a low viscosity formulation
- Two-part epoxy adhesive is highly compatible with many different materials
- Formulated with low viscosity for easy dispensing
Scotch-Weld™ Epoxy Adhesive DP190 adheres with tough, strong bonds and offers an extended work life. The adhesive is designed with a low viscosity formulation to give additional time for adjustment and make application easy.