Catalog # HERA1950842
Supplier: Thermo Scientific
Specifications
- Code couleur:argenté
- Pour:Thermo Scientific Matrix 200 µl Screwtop universal tubes, 500 µl storage tubes, 500 µl storage tubes with DuraSeal, 0,5 ml Nunc cryobank tubes, Thermo Scientific CryoExtra 94 (holds 92 racks)
- l×p×h ext.:135×930×838 mm
- Désignation:Rectanglular racks for tubes (cryogenic and ultra-low chest freezers), holds plates (22/rack)
- Cdt:1
Specifications
About this item
A convenient, streamlined process to maximize sample storage in your laboratory.
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200 up to 5,0 μl, offering maximised sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.