Anti-HDHD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Codice catalogo BOSSBS-8317R-A680
Nome del fornitore: Bioss
Specifiche
- Type d'anticorps:Primaire
- Nom de l'antigène:HDHD2
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Épitope:1-100/259
- Formulaire:Liquid
- ID de gène:84064
- Synonymes antigène:HDHD2_HUMAN|RGD1308579|Immediate early response 3 interacting protein 1|HDHD2|Haloacid dehalogenase-like hydrolase domain-containing protein 2|Ier3ip1|Haloacid dehalogenase like hydrolase domain containing 2|LRRG00122
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:29 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human HDHD2
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Specifiche
Informazioni su questo articolo
HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2) is also known as DKFZp564D1378 and is a 259 amino acid protein that is expressed as two isoforms produced by alternative splicing. HDHD2 belongs to the HAD-like hydrolase superfamily, which contains a group of hydrolase enzymes that differ from the å/
Type: Primary
Antigen: HDHD2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/259
Host: Rabbit
Isotype: IgG
Reactivity: