Quick-DNA™ Tissue/Insect Kits
À propos de cet article
Simple and efficient isolation of DNA from insects, including mosquitoes, bees, lice, ticks and D. melanogaster.
- Also compatible with tough-to-lyse tissues from other organisms
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
The Quick-DNA™ Tissue/Insect Kits are designed for the simple and rapid isolation of DNA (e.g., genomic, viral, mitochondrial) from fresh, frozen, or stored insect specimens - including mosquitoes, bees, lice, ticks, and D. melanogaster. The procedures are easy and can be completed in minutes. All kits are compatible with mammalian tissues, whole blood, and cultured cells. Samples are rapidly and efficiently lysed by bead beating with our state-ofthe- art, ultra-high density BashingBeads™. The DNA is then isolated and purified using our Zymo-Spin™ technology and is ideal for downstream molecular-based applications including PCR, array, genotyping, etc.
3 variantes
Product Details & Documents
Simple and efficient isolation of DNA from insects, including mosquitoes, bees, lice, ticks and D. melanogaster.
- Also compatible with tough-to-lyse tissues from other organisms
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
The Quick-DNA™ Tissue/Insect Kits are designed for the simple and rapid isolation of DNA (e.g., genomic, viral, mitochondrial) from fresh, frozen, or stored insect specimens - including mosquitoes, bees, lice, ticks, and D. melanogaster. The procedures are easy and can be completed in minutes. All kits are compatible with mammalian tissues, whole blood, and cultured cells. Samples are rapidly and efficiently lysed by bead beating with our state-ofthe- art, ultra-high density BashingBeads™. The DNA is then isolated and purified using our Zymo-Spin™ technology and is ideal for downstream molecular-based applications including PCR, array, genotyping, etc.
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- ZYMOD6015
- ZYMOD6016
- ZYMOD6017
- Désignation:
- Quick-DNA™ tissue/insect microprep kit, 50 preps
- Quick-DNA™ tissue/insect microprep kit, 50 preps
- Quick-DNA™ tissue/insect microprep kit, 2×96 preps
- Target molecule:
- DNA
- DNA
- DNA
- Refine target molecule:
- Total DNA
- Total DNA
- Total DNA
- Sample type:
- Cultured cells|Invertebrates|Human/animal cells and tissues
- Cultured cells|Invertebrates|Human/animal cells and tissues
- Cultured cells|Invertebrates|Human/animal cells and tissues
- Method / Format:
- Column (silica-based)
- Column (silica-based)
- Multiwell plate
- Prep size:
- Mini
- Mini
- Mini
Spécifications
Product Family Options
Informations sur le produit
- DésignationFormatCdtDisponibilitéPrix
- Quick-DNA™ tissue/insect microprep kit, 50 prepsSpin column1 KITConnexion Pour la disponibilité1 * 1 KITSFr. 258.00Spécifications :Marketsource
Plus
Specifications
Cat. No.ZYMOD6015DésignationQuick-DNA™ tissue/insect microprep kit, 50 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeCultured cells|Invertebrates|Human/animal cells and tissuesMethod / FormatColumn (silica-based)Prep sizeMiniMarketsource - Quick-DNA™ tissue/insect microprep kit, 50 prepsSpin column1 KITConnexion Pour la disponibilité1 * 1 KITSFr. 258.00Spécifications :Marketsource
Plus
Specifications
Cat. No.ZYMOD6016DésignationQuick-DNA™ tissue/insect microprep kit, 50 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeCultured cells|Invertebrates|Human/animal cells and tissuesMethod / FormatColumn (silica-based)Prep sizeMiniMarketsource - Quick-DNA™ tissue/insect microprep kit, 2×96 preps96 well spin column plate2 KITConnexion Pour la disponibilité1 * 2 KITSFr. 868.00Spécifications :Marketsource
Plus
Specifications
Cat. No.ZYMOD6017DésignationQuick-DNA™ tissue/insect microprep kit, 2×96 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeCultured cells|Invertebrates|Human/animal cells and tissuesMethod / FormatMultiwell platePrep sizeMiniMarketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



