Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Quick-DNA™ Kits

Quick-DNA™ Kits

Marketsource
Fournisseur:   Zymo Research
Quick-DNA™ Kits
Fournisseur:  Zymo Research
Marketsource
Certificats
Quick-DNA™ Kits

7 variantes

Product Details & Documents

DNA from cells, swabs, and whole blood.

  • Quick & easy: Simple 20 minute procedure
  • Unique technology excludes the use of proteinase K and organic denaturants
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ kits are ideal for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin™ column/plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.

Spécifications de la famille de produits

Spécifications

Spécifications

  • Catalog No:
  • ZYMOD3020
  • ZYMOD3021
  • ZYMOD3024
  • ZYMOD3025
  • ZYMOD3010
  • ZYMOD3011
  • ZYMOD3012
  • Désignation:
  • Quick-DNA™ Microprep kit, 50 preps
  • Quick-DNA™ Microprep kit, 200 preps
  • Quick-DNA™ Miniprep kit, 50 preps
  • Quick-DNA™ Miniprep kit, 200 preps
  • Quick-DNA™ 96 kit, 2×96 preps
  • Quick-DNA™ 96 kit, 4×96 preps
  • Quick-DNA™ 96 kit, 10×96 preps
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • DNA
  • DNA
  • DNA
  • DNA
  • Refine target molecule:
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Sample type:
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Method / Format:
  • Column (silica-based)
  • Column (silica-based)
  • Column (silica-based)
  • Column (silica-based)
  • Multiwell plate
  • Multiwell plate
  • Multiwell plate
  • Prep size:
  • Micro
  • Micro
  • Mini
  • Mini
  • Mini
  • Mini
  • Mini
  • Taille de l'échantillon:
  • ≤1×10⁶ cells or 5 mg tissue
  • ≤1×10⁶ cells or 5 mg tissue
  • ≤5×10⁶ cells or 25 mg tissue
  • ≤5×10⁶ cells or 25 mg tissue
  • ≤1×10⁶ cells or 5 mg tissue
  • ≤1×10⁶ cells or 5 mg tissue
  • ≤1×10⁶ cells or 5 mg tissue
  • Capacité d'échantillons:
  • 5 µg
  • 5 µg
  • 25 µg
  • 25 µg
  • 5 µg
  • 5 µg
  • 5 µg
  • Yield:
  • Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 25 µg/well total DNA is eluted into ≥50 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 25 µg/well total DNA is eluted into ≥50 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.

Spécifications

Product Family Options

Informations sur le produit

  • Désignation
    Cdt
    Disponibilité
    Prix
  • Quick-DNA™ Microprep kit, 50 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 150.00 SFr.
    150.00 SFr.
    1 * 1 KIT
       150.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3020
    Désignation
    Quick-DNA™ Microprep kit, 50 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Micro
    Taille de l'échantillon
    ≤1×10⁶ cells or 5 mg tissue
    Capacité d'échantillons
    5 µg
    Yield
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ Microprep kit, 200 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 489.00 SFr.
    489.00 SFr.
    1 * 1 KIT
       489.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3021
    Désignation
    Quick-DNA™ Microprep kit, 200 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Micro
    Taille de l'échantillon
    ≤1×10⁶ cells or 5 mg tissue
    Capacité d'échantillons
    5 µg
    Yield
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ Miniprep kit, 50 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 150.00 SFr.
    150.00 SFr.
    1 * 1 KIT
       150.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3024
    Désignation
    Quick-DNA™ Miniprep kit, 50 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Taille de l'échantillon
    ≤5×10⁶ cells or 25 mg tissue
    Capacité d'échantillons
    25 µg
    Yield
    Up to 25 µg/well total DNA is eluted into ≥50 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ Miniprep kit, 200 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 489.00 SFr.
    489.00 SFr.
    1 * 1 KIT
       489.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3025
    Désignation
    Quick-DNA™ Miniprep kit, 200 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Taille de l'échantillon
    ≤5×10⁶ cells or 25 mg tissue
    Capacité d'échantillons
    25 µg
    Yield
    Up to 25 µg/well total DNA is eluted into ≥50 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ 96 kit, 2×96 preps
    2 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 2 KIT
    1 * 2 KIT - 327.00 SFr.
    327.00 SFr.
    1 * 2 KIT
       327.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3010
    Désignation
    Quick-DNA™ 96 kit, 2×96 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Multiwell plate
    Prep size
    Mini
    Taille de l'échantillon
    ≤1×10⁶ cells or 5 mg tissue
    Capacité d'échantillons
    5 µg
    Yield
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ 96 kit, 4×96 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 614.00 SFr.
    614.00 SFr.
    1 * 1 KIT
       614.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3011
    Désignation
    Quick-DNA™ 96 kit, 4×96 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Multiwell plate
    Prep size
    Mini
    Taille de l'échantillon
    ≤1×10⁶ cells or 5 mg tissue
    Capacité d'échantillons
    5 µg
    Yield
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
  • Quick-DNA™ 96 kit, 10×96 preps
    1 KIT
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 KIT
    1 * 1 KIT - 1 311.00 SFr.
    1 311.00 SFr.
    1 * 1 KIT
       1 311.00 SFr.

    Spécifications :
    Marketsource
    Plus
    Specifications
    Cat. No.
    ZYMOD3012
    Désignation
    Quick-DNA™ 96 kit, 10×96 preps
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Multiwell plate
    Prep size
    Mini
    Taille de l'échantillon
    ≤1×10⁶ cells or 5 mg tissue
    Capacité d'échantillons
    5 µg
    Yield
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.