Mix & Go E. coli Transformation Kit and Buffer Set
Mix & Go E. coli transformation kit, 1 KIT
N° de catalogue ZYMOT3001
Marketsource
Fournisseur: Zymo Research
Mix & Go E. coli Transformation Kit and Buffer Set
N° de catalogue ZYMOT3001
Fournisseur: Zymo Research
Marketsource
Numéro CAS:
Aller à
Product Details & Documents
The Mix & Go kit convenient for the preparation of competent E. coli.
- It is very simple and efficient DNA transformation
- It completely eliminates the requirement for heat shocking and related procedures
- Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA
The Mix & Go E. coli transformation kit includes all buffers and ZymoBroth™ medium to generate 20 ml of Mix & Go competent cells.
Spécifications
Spécifications
- Cdt:1 KIT
- Désignation:Mix & Go E. coli transformation kit