Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant GROalpha/CXCL1 (from E. coli)
Human Recombinant GROalpha/CXCL1 (from E. coli)
Catalog # PEPR100-32-5UG
Supplier:  PeproTech, Inc.
CAS Number:  
undefined
Human Recombinant GROalpha/CXCL1 (from E. coli)
Catalog # PEPR100-32-5UG
Supplier:  PeproTech, Inc.
CAS Number:  

Specifications

  • Cdt:
    5 µG
  • Fonction des protéines:
    Chemokine
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at –20 °C prior to opening
  • Teneur en endotoxines:
    ≤1,00 EUs/μg
  • Activité biologique:
    Bioactive, but no test is suitable to set an Acceptance Criteria (primary cell assay, in vivo assay or cell line is not commercially available)
  • Reconstitution instructions:
    Reconstitute with Sterile water at 0,1 mg/ml
  • Synonyme de protéine:
    CXCL1|mKC|GRO1|MGSAα|NAP-3|rCINC
  • Base de données UnitProt:
    P09341
  • Nom de la protéine/du peptide:
    GRO alpha
  • Purity:
    >95%
  • Molecular weight:
    7,9 (kDa) Monomer
  • Sequence:
    ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN
  • Niveau d'endotoxine:
    Low
  • Formulation:
    Lyophilised from a sterile (0,2 µm) filtered aqueous solution containing 0,1% Trifluoroacetic Acid (TFA)
  • Température de transport:
    Ambient

Specifications

About this item

Growth regulated protein alpha (GRO-alpha), also known as CXCL1, is a chemokine that has mitogenic properties and is a neutrophil chemoattractant. GRO-alpha is secreted by macrophages, epithelial cells, neutrophils, and melanomas. GRO-alpha signals through the CXCR2 chemokine receptor and is important during spinal cord formation, inflammation, angiogenesis, tumorigenesis, and wound healing. 

  • USA origin

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.

Ordering information: Ships at room temperature; Store at –20 °C upon delivery.