Attachment Factor™ Extracellular Matrix
Attachment Factor™ Extracellular Matrix
Marketsource
Fournisseur:   Chemicals
Attachment Factor™ Extracellular Matrix
Fournisseur:  Chemicals
Marketsource
Certificats
Attachment Factor™ Extracellular Matrix
Attachment Factor™ Extracellular Matrix
2 variantes
Product Details & Documents
Promotes cell attachment to tissue culture surfaces to improve plating efficiency and encourage healthy growth.
  • High quality specialised reagents
  • Always available
  • Overnight delivery
Attachment Factor™ is an extracellular matrix (ECM) product that promotes cell attachment to tissue culture surfaces and encourages correct polarity and cytoskeletal organisation. Use of Attachment Factor™ is critically important when cultures are to be initiated, grown, passaged, or used within the Cell Systems Media family.
The biologically active elements of Attachment Factor™ are derived by a chromatographic process from heat-inactivated charcoal-stripped fetal bovine serum, and delivered at 1X in a PBS-based HEPES-buffered gelatin vehicle. Cell Systems exclusively uses and strongly recommends the use of serum products 100% traceable to cattle processed in the US at USDA inspected facilities.
Use of Attachment Factor™ improves plating efficiency, shortens 'lag time' after passage, enhances growth rate, and promotes the organisation of polarity and cytoskeletal architecture, and can dramatically increase cell yield during cell culture expansion. Use with all cultures except Melanocyte and Keratinocyte cells.
Handling and Storage: Store Attachment Factor™ at <−20 °C. Once product is opened, shelf life is 30 days at refrigerated temperatures (2 to 8 °C). Warm to 37 °C in water bath before each use. Refrigerate immediately after use.
Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.
Product Family Options
Informations sur le produit
  • Désignation
    Cdt
    Disponibilité
    Prix
  • Reagent
    10 mL
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 10 mL
    1 * 10 mL - SFr. 50.12
    SFr. 50.12
    1 * 10 mL
       SFr. 50.12
    Spécifications :
    Marketsource
    Marketsource
  • Reagent
    100 ml
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ml
    1 * 100 ml
This product has been blocked by your organization. Please contact your purchasing department for more information