Quick-RNA™ Plus Kits
À propos de cet article
RNA from any sample (cells, tissues, biological fluids and more).
- DNA-free: Genomic DNA removal column and DNase I included
- High quality: Ensure intact RNA with DNA/RNA Shield™ for sample storage at ambient temperatures
- NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more
The Quick-RNA™ miniprep plus kit is an innovative and versatile product designed for the easy, reliable, and rapid isolation of DNA-free RNA from cells, all tissue types, whole blood, and biological fluids. The provided DNA/RNA Shield™ stabilises samples, allowing them to be stored without the need for immediate freesing or processing for up to one month. Furthermore, DNA/RNA Shield™ inactivates RNases as well as microbial pathogens (viruses, bacteria and more). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17 to 200 nt).
Simply add DNA/RNA Shield™ and proteinase K to extract total RNA from any tissue, then purify the RNA using the Zymo-Spin™ column. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more. In addition, the kit can be used for the enrichment of small and large RNAs in two separate fractions.
2 variantes
Product Details & Documents
RNA from any sample (cells, tissues, biological fluids and more).
- DNA-free: Genomic DNA removal column and DNase I included
- High quality: Ensure intact RNA with DNA/RNA Shield™ for sample storage at ambient temperatures
- NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more
The Quick-RNA™ miniprep plus kit is an innovative and versatile product designed for the easy, reliable, and rapid isolation of DNA-free RNA from cells, all tissue types, whole blood, and biological fluids. The provided DNA/RNA Shield™ stabilises samples, allowing them to be stored without the need for immediate freesing or processing for up to one month. Furthermore, DNA/RNA Shield™ inactivates RNases as well as microbial pathogens (viruses, bacteria and more). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17 to 200 nt).
Simply add DNA/RNA Shield™ and proteinase K to extract total RNA from any tissue, then purify the RNA using the Zymo-Spin™ column. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more. In addition, the kit can be used for the enrichment of small and large RNAs in two separate fractions.
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- ZYMOR1057
- ZYMOR1058
- Target molecule:
- RNA
- RNA
- Refine target molecule:
- Total RNA
- Total RNA
- Sample type:
- Cultured cells|Human/animal cells and tissues
- Cultured cells|Human/animal cells and tissues
- Method / Format:
- Column (silica-based)
- Column (silica-based)
- Prep size:
- Mini
- Mini
- Taille de l'échantillon:
- ≤1×10⁷ Cells or 50 mg Tissue
- ≤1×10⁷ Cells or 50 mg Tissue
- Capacité d'échantillons:
- 100 µg
- 100 µg
- Yield:
- spin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)
- spin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)
Spécifications
Product Family Options
Informations sur le produit
- DésignationCdtDisponibilitéPrix
- Spécifications :Marketsource
Plus
Specifications
Cat. No.ZYMOR1057Target moleculeRNARefine target moleculeTotal RNASample typeCultured cells|Human/animal cells and tissuesMethod / FormatColumn (silica-based)Prep sizeMiniTaille de l'échantillon≤1×10⁷ Cells or 50 mg TissueCapacité d'échantillons100 µgYieldspin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)Marketsource - Spécifications :Marketsource
Plus
Specifications
Cat. No.ZYMOR1058Target moleculeRNARefine target moleculeTotal RNASample typeCultured cells|Human/animal cells and tissuesMethod / FormatColumn (silica-based)Prep sizeMiniTaille de l'échantillon≤1×10⁷ Cells or 50 mg TissueCapacité d'échantillons100 µgYieldspin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)Marketsource
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



