Zymo-Spin™ Column/Filter Assemblies
About this item
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
- For purifying DNA and or/RNA from high-volume sample inputs
- High-recovery
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
8 Options Available Below
Product Details & Documents
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
- For purifying DNA and or/RNA from high-volume sample inputs
- High-recovery
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
Product Family Options
Product Information
- DésignationFormatCdtAvailabilityPrice
- Zymo-Spin™ VI with Zymo-Maxi Filter™Spin column/filter assembly10Log in for availability1 * 10 STSFr. 81.00Specifications:MarketsourceMarketsource
- Zymo-Spin™ VI with ReservoirSpin column/filter assembly10Log in for availability1 * 10 STSFr. 77.00Specifications:MarketsourceMarketsource
- Zymo-Spin™ VI with Zymo-Maxi Filter™Spin column/filter assembly20Log in for availability1 * 20 STSFr. 127.00Specifications:MarketsourceMarketsource
- Zymo-Spin™ VI with ReservoirSpin column/filter assembly20Log in for availability1 * 20 STSFr. 121.00Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Zymo-Spin™ V with reservoirSpin column/filter assembly25Log in for availability1 * 25 STSFr. 97.00Specifications:MarketsourceMarketsource
- Zymo-Spin™ V with reservoirSpin column/filter assembly50Log in for availability1 * 50 STSFr. 152.00Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource