Zymo-Spin™ Column/Filter Assemblies
Marketsource
Supplier:   Zymo Research
Zymo-Spin™ Column/Filter Assemblies
Supplier:  Zymo Research
Marketsource
Certificates
Zymo-Spin™ Column/Filter Assemblies
Zymo-Spin™ Column/Filter Assemblies
8 Options Available Below
Product Details & Documents
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
  • For purifying DNA and or/RNA from high-volume sample inputs
  • High-recovery
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
Product Family Options
Product Information
  • Désignation
    Format
    Cdt
    Availability
    Price
  • Zymo-Spin™ VI with Zymo-Maxi Filter™
    Spin column/filter assembly
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 81.00
    SFr. 81.00
    1 * 10 ST
       SFr. 81.00
    Specifications:
    Marketsource
    Marketsource
  • Zymo-Spin™ VI with Reservoir
    Spin column/filter assembly
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 77.00
    SFr. 77.00
    1 * 10 ST
       SFr. 77.00
    Specifications:
    Marketsource
    Marketsource
  • Zymo-Spin™ VI with Zymo-Maxi Filter™
    Spin column/filter assembly
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 127.00
    SFr. 127.00
    1 * 20 ST
       SFr. 127.00
    Specifications:
    Marketsource
    Marketsource
  • Zymo-Spin™ VI with Reservoir
    Spin column/filter assembly
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 121.00
    SFr. 121.00
    1 * 20 ST
       SFr. 121.00
    Specifications:
    Marketsource
    Marketsource
  • ZRC-GF Filter™
    Spin column/filter assembly
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 91.00
    SFr. 91.00
    1 * 20 ST
       SFr. 91.00
    Specifications:
    Marketsource
    Marketsource
  • Zymo-Spin™ V with reservoir
    Spin column/filter assembly
    25
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 97.00
    SFr. 97.00
    1 * 25 ST
       SFr. 97.00
    Specifications:
    Marketsource
    Marketsource
  • Zymo-Spin™ V with reservoir
    Spin column/filter assembly
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 152.00
    SFr. 152.00
    1 * 50 ST
       SFr. 152.00
    Specifications:
    Marketsource
    Marketsource
  • ZRC-GF Filter™
    Spin column/filter assembly
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 216.00
    SFr. 216.00
    1 * 50 ST
       SFr. 216.00
    Specifications:
    Marketsource
    Marketsource