Malachite G Phosphate Assay Kit
Catalog # MSPPPOMG-25H
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Phosphate assay kit
- Method:OD620 nm (malachite green).
- Sample:Biological, environment etc.
Specifications
About this item
For sensitive and high-throughput phosphate determination.
- Very stable reagent
- High sensitivity and wide detection range (0,02 to 40 μM phosphate)
- Homogeneous 'mix-and-measure' type high-throughput assay
- Assays can be performed in tubes, cuvettes or microplates
The Malachite Green Phosphate Assay Kit is based on quantification of the green complex formed between Malachite Green, molybdate and free orthophosphate. The rapid colour formation from the reaction can be conveniently measured on a spectrophotometer (600 to 660 nm) or on a plate reader. The non-radioactive colorimetric assay kits have been optimized to offer superior sensitivity and prolonged. Shelf Life. The assay is simple and fast, involving a single addition step for phosphate determination. Assays can be executed in tubes, cuvettes or multi-well plates. The assays can be conveniently performed in 96- and 384-well plates for high-throughput screening of enzyme inhibitors.