Thermo Scientific™ Remel™ Agglutinating Sera
Remel Agglutinating Sera are used for serological identification of Bordetella cultures by rapid slide agglutination test.
- Température de stockage:2 °C to 8 °C
Frequently Bought Together
Remel Agglutinating Sera are used for serological identification of Bordetella cultures by rapid slide agglutination test.