Pouch masks with nylon headband, non sterile, HM4, PUREZERO®
N° de catalogue 114-9601
Fournisseur: Halyard
Spécifications
- Type:Pouch masks with headband, non sterile
- Couleur:blanc
- Cdt:300
Spécifications
À propos de cet article
The HM4 Pouch Mask is a three-layer mask with nylon headband with an expanded breathing chamber that provides high particle, bacterial, and viral filtration efficiency. The mask construction consists of low-linting materials and a polyester foam nose band.
- Three-layer mask
- Nylon headband
- Designed for industrial and cleanroom use
- Recommended for ISO Class 4 or higher and Grade B/C/D cleanrooms
- Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the task with PUREZERO® HM4 non-sterile face masks, which are specifically designed to meet the stringent requirements of cleanroom environments. This three-layer mask features a unique bicomponent material (polyester and polyethylene). They are manufactured in facilities with raw materials and components traceable to the original supplier.