Cleanroom sleeves, CleanMax®
Catalog # 114-9544
Supplier: LAKELAND CLEANMAX APPAREL
Specifications
- Applications des tenues:Salle propre
- Couleur:blanc
- Longueur:18"
- Cdt:50 PAIR
- Classe de la salle propre:100/ISO 5
Specifications
About this item
Clean manufactured nonsterile sleeves with bound seams. Clean manufactured in a silicone-free environment.
- Fluid repellent; tested in accordance with ASTM D6978 for chemotherapy permeation
- Open fold for easier donning
- Resistant to blood, body fluid and viral penetration
- Resistant to blood borne pathogens
- IEST-RP-CC003 Category I particle cleanliness
- Latex-free and silicone-free
CleanMax® cleanroom garments for controlled environments provide optimal comfort, quality, and protection. Ideally suited for use in ISO Class 4 - 8 cleanrooms and all controlled environments. CleanMax® garments are easy to don, comfortable to wear, and are backed by Lakeland's 30+ years as a manufacturer of disposable protective apparel.
Packaging: Packaged in pairs in a protective outer bag for ante areas.