Order Entry
Switzerland
ContactUsLinkComponent
16Alpha-Hydroxy Prednisolone-9(11)-ene Acetate, Pharmaceutical Analytical Impurities
16Alpha-Hydroxy Prednisolone-9(11)-ene Acetate, Pharmaceutical Analytical Impurities
N° de catalogue 1A01270.
Fournisseur:  USP
Numéro CAS:  
16Alpha-Hydroxy Prednisolone-9(11)-ene Acetate, Pharmaceutical Analytical Impurities
N° de catalogue 1A01270.
Fournisseur:  USP
Numéro CAS:  

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    25 mg
  • Normes:
    Pharmaceutical Analytical Impurities
  • N ° CAS.:
    77017-20-0
  • Désignation:
    16Alpha-Hydroxy Prednisolone-9(11)-ene Acetate
  • Synonyme(s):
    2-((8S,10S,13S,14S,16R,17S)-16,17-dihydroxy-10,13-dimethyl-3-oxo-6,7,8,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl acetate
  • Désignation:
    16Alpha-Hydroxy Prednisolone-9(11)-ene Acetate

Spécifications

À propos de cet article

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.