16Alpha-Hydroxy Prednisolone-14-ene Acetate, Pharmaceutical Analytical Impurities
Catalog # 1A01220.
Supplier: USP
CAS Number:
Specifications
- Type de conditionnement:Vial
- Cdt:25 mg
- Normes:Pharmaceutical Analytical Impurities
- N ° CAS.:131918-72-4
- Désignation:16Alpha-Hydroxy Prednisolone-14-ene Acetate
- Synonyme(s):2-Oxo-2-((8R,9S,10R,11S,13S,16R,17S)-11,16,17-trihydroxy-10,13-dimethyl-3-oxo-6,7,8,9,10,11,12,13,16,17-decahydro-3H-cyclopenta[a]phenanthren-17-yl)ethyl acetate
- Désignation:16Alpha-Hydroxy Prednisolone-14-ene Acetate
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.