Order Entry
Switzerland
ContactUsLinkComponent
tert-Octylamine, Pharmaceutical Analytical Impurities
  1A01310.
 :  USP
 :  
tert-Octylamine, Pharmaceutical Analytical Impurities
  1A01310.
 :  USP
 :  

 

  • Type de conditionnement:
    Ampule
  • Cdt:
    25 mg
  • Normes:
    Pharmaceutical Analytical Impurities
  • N ° CAS.:
    107-45-9
  • Désignation:
    tert-Octylamine
  • UN No.:
    2734
  • Synonyme(s):
    2,4,4-trimethylpentan-2-amine
  • Désignation:
    tert-Octylamine

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.