Order Entry
Switzerland
ContactUsLinkComponent
Chloro N-methylthiophene Carboxamide, Pharmaceutical Analytical Impurities
Chloro N-methylthiophene Carboxamide, Pharmaceutical Analytical Impurities
Catalog # 1A01450.
Supplier:  USP
CAS Number:  
Chloro N-methylthiophene Carboxamide, Pharmaceutical Analytical Impurities
Catalog # 1A01450.
Supplier:  USP
CAS Number:  

Specifications

  • Type de conditionnement:
    Vial
  • Cdt:
    25 mg
  • Normes:
    Pharmaceutical Analytical Impurities
  • N ° CAS.:
    97799-98-9
  • Désignation:
    Chloro N-methylthiophene Carboxamide
  • Synonyme(s):
    5-chloro-N-methylthiophene-2-carboxamide
  • Désignation:
    Chloro N-methylthiophene Carboxamide

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.