Alpha-Naphthol, Pharmaceutical Analytical Impurities
Catalog # 1A01070.
Anbieter: USP
CAS-Nummer:
Technische Daten
- Type de conditionnement:Vial
- Cdt:25 mg
- Normes:Pharmaceutical Analytical Impurities
- N ° CAS.:90-15-3
- Désignation:Alpha-Naphthol
- UN No.:2811
- Synonyme(s):Naphthalen-1-ol
- Désignation:Alpha-Naphthol
Technische Daten
Über diesen Artikel
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.