Order Entry
Switzerland
ContactUsLinkComponent
3'-(N,N-Didemethyl)azithromycin;, Pharmaceutical Analytical Impurities
3'-(N,N-Didemethyl)azithromycin;, Pharmaceutical Analytical Impurities
N° de catalogue 1A00070.
Fournisseur:  USP
Numéro CAS:  
3'-(N,N-Didemethyl)azithromycin;, Pharmaceutical Analytical Impurities
N° de catalogue 1A00070.
Fournisseur:  USP
Numéro CAS:  

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    25 mg
  • Normes:
    Pharmaceutical Analytical Impurities
  • N ° CAS.:
    612069-27-9
  • Désignation:
    3'-(N,N-Didemethyl)azithromycin;
  • Synonyme(s):
    Aminoazithromycin
  • Désignation:
    3'-(N,N-Didemethyl)azithromycin;

Spécifications

À propos de cet article

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.