17-Oxo Dexamethasone, Pharmaceutical Analytical Impurities
N° de catalogue 1A00470.
Fournisseur: USP
Numéro CAS:
Spécifications
- Type de conditionnement:Vial
- Cdt:10 mg
- Normes:Pharmaceutical Analytical Impurities
- N ° CAS.:1880-61-1
- Désignation:17-Oxo Dexamethasone
- Synonyme(s):(8S,9R,10S,11S,13S,14S,16R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-7,8,9,10,11,12,13,14,15,16-decahydro-3H-cyclopenta[a]phenanthrene-3,17(6H)-dione
- Désignation:17-Oxo Dexamethasone
Spécifications
À propos de cet article
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.